5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: NHS-Fluorescein, FLUOS, 5(6)-FAM SE

112 110 111
AATB112EA 1480 EUR
AATB112 AATB110 AATB111
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥95%
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)

Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.

Formel: C₂₅H₁₅NO₉
MDL Number: MFCD04119713
CAS-Nummer: 117548-22-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2bacf63d577b79d45a3dc6ce66aa7a92dd0d74&vid=a43d34e7c9bf4144e1da9e7dc01ab624c925f849&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR