5-Carboxyfluorescein ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate
103
105
104
AATB103EA
120
EUR
AATB103
AATB105
AATB104
5-Carboxyfluorescein ≥95%
5-Carboxyfluorescein
5-FAM is the purified single isomer of carboxyfluorescein.
Formel:
C₂₁H₁₂O₇.xH₂O |
MDL Number:
MFCD00036874 CAS-Nummer: 76823-03-5 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...