Human Protein A-Agarose Resin
Lieferant: AAT Bioquest
55000 55005
AATB55000EA 152 EUR
AATB55000 AATB55005
Human Protein A-Agarose Resin
Proteine und Peptide
Protein A agarose was prepared by covalently immobilizing protein A onto high-quality crosslinked beaded agarose.
  • High affinity
  • Targets igg antibodies
  • pH stable between 3 to 13
Bitte beachten For research use only.