7-Amino-4-(trifluormethyl)cumarin ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
7-Amino-4-(trifluoromethyl)coumarin, 7-Amino-4-trifluoromethylcoumarin, 7-Amino-4-(trifluoromethyl)-2H-chromen-2-one, Coumarin 151
56
57
58
AATB56EA
152
EUR
AATB56
AATB57
AATB58
7-Amino-4-(trifluormethyl)cumarin ≥95%
7-Amino-4-(trifluormethyl)cumarin
This compound is the precursor for a variety of AFC enzyme substrates.
Formel:
C₁₀H₆F₃NO₂ |
MDL Number:
MFCD00006858 CAS-Nummer: 53518-15-3 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...