Fmoc-Lys(Dabcyl)-OH ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nα-Fmoc-L-lysine

5040 5041
AATB5040EA 152 EUR
AATB5040 AATB5041
Fmoc-Lys(Dabcyl)-OH ≥90%
Fmoc-Lys(Dabcyl)-OH

FMOC-Lys(DABCYL)-OH is a building block for in-sequence Lys labeling by DABCYL.

Formel: C₃₆H₃₇N₅O₅
Molekulargewicht: 619.722 g/mol
MDL Number: MFCD00467694
CAS-Nummer: 146998-27-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1477885921ad694d8dabba053c13991781862b6c&vid=b0fb7ea94e726f9d83f44f136859b1b812ad5de3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR