FMOC-Asp(biotinyl-PEG)-OH
Lieferant: AAT BIOQUEST
Achtung
5023 5024
AATB5023EA 307 EUR
AATB5023 AATB5024
FMOC-Asp(biotinyl-PEG)-OH
FMOC-Asp(biotinyl-PEG)-OH
This PEG biotin-labeled amino acid has excellent solubility in DMF and other solvents used in SPPS.
  • Specific reactivity
  • Stable conjugation
  • High solubility
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a74991d588a8d5241eca8a0aaf1dad4e591ae4e&vid=5973d07e03e9b332fd5d9e4f97c7f480f99d03f2&prid=web&clid=SAPDAM