6-Carboxyfluorescein ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

106 107 108
AATB106EA 120 EUR
AATB106 AATB107 AATB108
6-Carboxyfluorescein ≥95%
6-Carboxyfluorescein

6-FAM is the other purified isomer of 5(6)-carboxyfluorescein.

Formel: C₂₁H₁₂O₇
MDL Number: MFCD00036873
CAS-Nummer: 3301-79-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd80d3ff1bb5b71ce65a794dee606f3900284a92&vid=3ab92665e6217afe078170a638027f190dd477bb&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR