VWR® Laboratory LED Zoom Stereo Series
Lieferant: VWR Collection
10836-004 10836-006
VWRU10836-004EA 0 EUR
VWRU10836-004 VWRU10836-006
VWR® Laboratory LED Zoom Stereo Series
Mikroskope
The Laboratory LED Zoom Stereo Series is a premium line of stereo microscopes offered by Avantor. Its quality construction and Greenough optical system make it a durable workhorse when dealing with all sorts of biological, medical and industrial applications. With a large working distance, and wide zoom range, the variety of applications for this microscope is limitless.
  • The Greenough Optical Zoom System ensures durability and consistent image quality over the long lifetime of the microscope
  • A large zoom range and working distance of 113 mm provides flexibility to properly examine all kinds of different samples
  • Rubber eye guards are included to minimize stray light
  • Distortion free images with high resolving power (220 lp/mm in standard configuration)
  • Achieves parfocality through the complete magnification range, without need to re-adjust focus position while zooming
  • Auxiliary objectives and eyepieces are available separately, to increase working distance or magnification range
  • Incident and transmitted LED light sources provide high quality lighting conditions and maximum flexibility for lighting samples
VWR® Laboratory LED Zoom Stereo Series is a premium line of stereo microscopes, designed to take on the demands of repeated professional laboratory and industrial applications
High quality optics add to this microscope’s flexibility. Its Widefield 10×/23 mm eyepieces and its 6.7:1 zoom ratio, enabling a magnification range of 0,75× to 5×, make this microscope more than capable of delivering vivid, detailed images of a wide variety of samples. With a high resolving power of 220 lp/mm (in standard configuration), users will never be left disappointed with their results.
The ergonomic design and usability of this microscope cannot be understated. With a working distance of 113 mm (extendable to 324 mm with a .3× auxiliary objective) and large working area stand, samples big and small can be scrutinized with ease. Standard eyepieces include rubber eyecups, which remove outside light, reducing eye strain. Eyepieces also include a diopter adjustment (±5 dpt) allowing those who wear glasses to see the entire image with ease. The available trinocular version makes this microscope perfect for photomicrography.
Those seeking a premium stereomicroscope for industrial quality control or biological preparation need not look any further than VWR® Laboratory LED Zoom Stereo Series. This microscope provides an incredible value and is an essential addition to any professional microscopist’s tool kit.