Safety Shoes, Clogs, Active, 87031
Lieferant: Abeba
87031/36 87031/37 87031/38 87031/39 87031/40 87031/41 87031/42 87031/43 87031/44 87031/45 87031/46 87031/47 87031/48 87031/35
ABEB87031/36EA 47.3 EUR
ABEB87031/36 ABEB87031/37 ABEB87031/38 ABEB87031/39 ABEB87031/40 ABEB87031/41 ABEB87031/42 ABEB87031/43 ABEB87031/44 ABEB87031/45 ABEB87031/46 ABEB87031/47 ABEB87031/48 ABEB87031/35
Safety Shoes, Clogs, Active, 87031
Schuhe und Stiefel
Anti-static safety clogs made from grained leather with a non foldable, adjustable heel strap and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Increased breathability
  • Increased durability of the upper
  • Easy-clean upper
  • Sole resistance to acids and bases
  • Adjustable instep strap with buckle fastening
  • Energy absorption in the heel area - energy absorbtion in the heel area
  • The sole resistant to diesel fuel - sole resistance to diesel fuel
Suitable for use in gastronomy/kitchen, horeca, hospitals, pharmaceuticals.
Colour: Black, Toe: Thermo plastic resistant to deformation, light, elastic.
Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol - slip resistance on ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol. Antistatic footwear - antistatic properties. Energy absorption in the heel area - energy absorbtion in the heel area. The sole resistant to diesel fuel - sole resistance to diesel fuel.
Zertifizierungen: EN ISO 20347:2012; 19/2022/PPE/1439/B edition 1; OB (Basic requirements met); SRC (Slip resistance on ceramic).