Reflexor® Comfort, 36813 ESD Occupational Shoes, Medical Clogs
Lieferant: Abeba
36813/41 36813/36 36813/37 36813/38 36813/39 36813/40 36813/42 36813/43
ABEB36813/41EA 126 EUR
ABEB36813/41 ABEB36813/36 ABEB36813/37 ABEB36813/38 ABEB36813/39 ABEB36813/40 ABEB36813/42 ABEB36813/43
Reflexor® Comfort, 36813 ESD Occupational Shoes, Medical Clogs
Schuhe und Stiefel
Occupational antistatic medical clog made from grain leather and microfiber inner lining with fabric insole for women.
  • Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol
  • ESD - protection against electrostatic discharge
  • Antistatic footwear
  • Energy absorption in the heel area
Weight of half a pair is 0,30 kg for size 37.
Zertifizierungen: EN ISO 20347:2012, OB, SRC.