High Adhesion Self-Laminating Vinyl Wire Wraps, B33
Lieferant: Brady
153252 153253 153259 153261 153265 153257 153264 153260 153263 153258 153262 153254
BRDY153252EA 93.79 EUR
BRDY153252 BRDY153253 BRDY153259 BRDY153261 BRDY153265 BRDY153257 BRDY153264 BRDY153260 BRDY153263 BRDY153258 BRDY153262 BRDY153254
High Adhesion Self-Laminating Vinyl Wire Wraps, B33
Etiketten
Keep your wire identification secure with this self-laminating marker with extra-strong adhesive.
  • The extra-strong adhesive makes these markers ideal for small-gauge wires
  • Features a white printable area and a clear tail that wraps around the marker
  • Tail serves as an overlaminate to protect the print
  • A ribbon is required to print this material
  • Label type: B-417
Ribbon series: B30 monocolour series
Shape: Rectangle
Label properties: Self-laminating and wrap around
Resistance properties: Chemical-resistant, humidity-resistant, oil-resistant, water-resistant, weather-resistant
Applications: Data and telecommunications labelling, electrical labelling, wire and cable labelling.
Zertifizierungen: UL approved.