DermiFlex Ultra Gloves
Lieferant: Portwest
A352G8RS A352G8RXXL
PRTWA352G8RSEA 59.3 EUR
PRTWA352G8RS PRTWA352G8RXXL
DermiFlex Ultra Gloves
Handschuhe
The DermiFlex Ultra provides additional protection against water, oil, and grease due to the additional ³/₄ dip compared to the A350.
  • Flexible sandy nitrile coating offers great grip in wet and dry conditions
  • ³/₄ dipped for increased protection
  • 15 gauge liner for extra dexterity
  • OEKO-TEX® STANDARD 100 approved
  • Lightweight and comfortable
Low linting construction for minimal contamination, with a breathable seamless liner. The super light 15 gauge nylon liner has a soft nitrile foam palm for the ultimate comfort, dexterity, and sensitivity. Retail tag aids presentation for retail sales.
Colour: Grey/black
Zertifizierungen: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4131X. ANSI/ISEA 105: 2016 CUT Level A1, ANSI/ISEA 105: 2016 ABRASION Level 4. CE certified.