Chart Recorder Options for ULT Freezers
Lieferant: Thermo Scientific
332166G01 332166G02 332791G01 332791G02
HERA332166G01EA 1810 EUR
HERA332166G01 HERA332166G02 HERA332791G01 HERA332791G02
Chart Recorder Options for ULT Freezers
Gefriergeräte Ultratiefkühlgeräte
Chart recorder options for ultra-low temperature chest freezers.
  • Ink or inkless, physical record keeping for temperature tracking
Chart recorders provide an uneditable, 21 CFR Part 11 compliant, historical temperature recording for documentation and tracking.
Bestellinformation: Add to ordering configuration with ULT chest freezer.