Accessories for Rainin Carousel Stand
Lieferant: RAININ
HU-A3 17006638
RAINHU-A3EA 0 EUR
RAINHU-A3 RAIN17006638
Accessories for Rainin Carousel Stand
Pipettenständer
Adapter for use with Rainin electronic and multichannel pipettes on Hang-Ups and Carousel stand. 3 adapters per pack.
  • Clips into Hang-Up or Carousel, allows E4XLS+ pipettes to use these stands
  • Quick fitting: No tools required
  • Supplied as a pack of three