Polyethylene glycol 4500 NF
Lieferant: Spectrum Chemical
Synonyme:
PEG 4,500
P1792-12KG
P1792-2.5KG
SPCMP1792-12KGEA
1060
EUR
SPCMP1792-12KG
SPCMP1792-2.5KG
Polyethylene glycol 4500 NF
Polyethylenglycol 4.000
Polyethylene Glycol 4500, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formel:
H(OCH₂CH₂)nOH Molekulargewicht: 4500 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00081839 CAS-Nummer: 25322-68-3 |
Spezifikation Testergebnisse
Molecular Weight | 4050-4950 |
Completeness and Color of Solution | To Pass Test |
Viscosity @98.9 C (210 F) +/-0.3 C | 140- 200 Cs |
pH of a 5% Solution @ 25°C | 4.5- 7.5 |
Residue On Ignition | max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...