Ferrous gluconate dihydrate ≥97,0% USP
Lieferant: Spectrum Chemical
Ferrous Gluconate, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₁₂H₂₂FeO₁₄.2H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150872 CAS-Nummer: 12389-15-0 |
Spezifikation Testergebnisse
Assay (Dried Basis) | 97.0 - 102.0% |
Loss On Drying | 6.5 - 10.0 % |
Chloride (Cl) | max. 0.07 % |
Sulfate | max. 0.1 % |
Oxalic Acid | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...