S-(Carboxymethyl)-L-cysteine
Lieferant: Spectrum Chemical
S-Carboxymethyl-L-cysteine, also known as Rhinathiol or Humex, is a mucolytic that minimizes the viscosity of sputum so it can be used to treat chronic obstructive pulmonary disorder and bronchiectasis.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₅H₉NO₄S Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00002614 CAS-Nummer: 638-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...