Peppermint oil NF

Lieferant: Spectrum Chemical

PE105-500MLGL PE105-100ML PE105-10KGBL
SPCMPE105-500MLGLEA 524 EUR
SPCMPE105-500MLGL SPCMPE105-100ML SPCMPE105-10KGBL
Peppermint oil NF
Peppermint oil

Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00147870
CAS-Nummer: 8006-90-4

Bestellen

Spezifikation Testergebnisse

Esters(As Menthyl Acetate) 5.0% min.
Total Menthol(Free and As Esters) 50.0 % min.
Specific Gravity @ 25°C 0.896 - 0.908
Angular Rotation -18° to -32°
Refractive Index @ 20°C 1.459 - 1.465


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR