Allantoin ≥98%, Pulver
Lieferant: Spectrum Chemical
Allantoin, Powder, also referred to as glyoxyldiureide, is an important metabolic intermediate in many mammals, plants and bacteria, exept humans and higher apes. It is present in botanical extracts and in urine from most animals. Synthesised Allantoin is nature-identical, safe and non-toxic and meets CTFA and JSCI requirements. It is used in toothpaste, mouthwash and other oral hygiene products.
Formel:
C₄H₆N₄O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005260 CAS-Nummer: 97-59-6 |
Spezifikation Testergebnisse
Assay | 98% min. |
Ash | max.0.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...