Carmine

Lieferant: Spectrum Chemical

Synonyme: Carminic acid, Cochineal, Alum lake of Carminic Acid, Natural Red 4, FD and C Carmine, CI 75470, Carmine, calcium/aluminum lake

CA195-1KGBL CA195-5KGBL
SPCMCA195-1KGBLEA 5510 EUR
SPCMCA195-1KGBL SPCMCA195-5KGBL
Carmine
Carmin (Calcium-Aluminium-Verlackung von Carminsäure)

Carmine, also known as cochineal or crimson lake, is a red colored pigment obtained from the aluminum salt of carminic acid. It is used in artificial flowers, paints and coloring. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₂₂H₂₀O₁₃
Molekulargewicht: 492.389 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00167028
CAS-Nummer: 1390-65-4

Bestellen

Spezifikation Testergebnisse

Absorption Max 531 (563) NM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR