Zinc sulphate heptahydrate ≥99%, körnig USP
Lieferant: Spectrum Chemical
|
Gefahr
|
ZI120-500GM
ZI120-12KG
ZI120-2.5KG
SPCMZI120-500GMEA
120
EUR
SPCMZI120-500GM
SPCMZI120-12KG
SPCMZI120-2.5KG
Zinc sulphate heptahydrate ≥99%, körnig USP
Zinksulfat Heptahydrat
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
ZnSO₄·7H₂O Molekulargewicht: 287.54 g/mol Schmelzpunkt: 100 °C Dichte: 1,97 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149894 CAS-Nummer: 7446-20-0 EINECS: 231-793-3 UN-Nummer: 3077 ADR: 9 III Merck Index: 12,10293 |
Spezifikation Testergebnisse
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...