Zinc stearate, Pulver USP
Lieferant: Spectrum Chemical
ZI115-500GM
ZI115-12KG
ZI115-2.5KG
SPCMZI115-500GMEA
142
EUR
SPCMZI115-500GM
SPCMZI115-12KG
SPCMZI115-2.5KG
Zinc stearate, Pulver USP
Zinkstearat
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₃₆H₇₀O₄Zn Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013031 CAS-Nummer: 557-05-1 |
|
Spezifikation Testergebnisse
Assay (ZnO) | 12.5 - 14.0% |
Alkalies and Alkaline Earths | max. 1.0 % |
Arsenic | max. 1.5 ppm |
Lead | max. 10 ppm |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...