Dimethyl phthalate ≥99%

Lieferant: Spectrum Chemical

Synonyme: Phthalic acid dimethyl ester

D1044-200LTBL D1044-20LT
SPCMD1044-200LTBLEA 3840 EUR
SPCMD1044-200LTBL SPCMD1044-20LT
Dimethyl phthalate ≥99%
Dimethylphthalat

Dimethyl Phthalate, an ectoparasiticide, is the methyl ester of phthalic acid and used in plastics, insect repellents, among other things.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₀H₁₀O₄
Siedepunkt: 283.7 °C (1013 hPa)
Schmelzpunkt: 6 °C
Dichte: 1,192 g/cm³ (20 °C)
Flammpunkt: 146 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008425
CAS-Nummer: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR