Dimethylammonium chloride

Lieferant: Spectrum Chemical

D2396-12KGBL D2396-2.5KGPL D2396-25KGBL
SPCMD2396-12KGBLEA 3080 EUR
SPCMD2396-12KGBL SPCMD2396-2.5KGPL SPCMD2396-25KGBL
Dimethylammonium chloride
Dimethylammoniumchlorid
Formel: C₂H₈ClN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012477
CAS-Nummer: 506-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR