Adenine hydrochloride

Lieferant: Spectrum Chemical

Synonyme: 7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride

A3192-1KGBL A3192-25KGBL
SPCMA3192-1KGBLEA 4800 EUR
SPCMA3192-1KGBL SPCMA3192-25KGBL
Adenine hydrochloride
Adenin Hydrochlorid
Formel: C₅H₅N₅·HCl
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150864
CAS-Nummer: 2922-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR