CBL0137 ≥95%

Lieferant: Cayman Chemical

Synonyme: 1,1'-[9-[2-[(1-methylethyl)amino]ethyl]-9H-carbazole-3,6-diyl]-bis-ethanone, 1,1'-(9-(2-(Isopropylamino)ethyl)-9H-carbazole-3,6-diyl)bis(ethan-1-one), Curaxin 137

19110-1 19110-10 19110-5
CAYM19110-1EA 63 EUR
CAYM19110-1 CAYM19110-10 CAYM19110-5
CBL0137 ≥95%
CBL0137

  • An activator of p53 and inhibitor of NF-κB


A metabolically stable curaxin that activates p53 with an EC₅₀ value of 0,37 µM and inhibits NF-κB with an EC₅₀ of 0,47 µM; has broad anticancer activity in mice when administered orally.

Formel: C₂₁H₂₄N₂O₂
Molekulargewicht: 336.4 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD18071578
CAS-Nummer: 1197996-80-7

Bestellen

Spezifikation Testergebnisse

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR