IKK Inhibitor VI ≥95%

Lieferant: Cayman Chemical

Synonyme: 5-Phenyl-2-ureidothiophene-3-carboxylic Acid Amide, 2-[(aminocarbonyl)amino]-5-phenyl-3-thiophenecarboxamide

17276-1 17276-500 17276-5
CAYM17276-1EA 148 EUR
CAYM17276-1 CAYM17276-500 CAYM17276-5
IKK Inhibitor VI ≥95%
IKK Inhibitor VI

  • An inhibitor of IKK2


A potent, cell-permeable, reversible inhibitor of IKK2 (IC₅₀= 13 nm); used to evaluate the role of the canonical, IκB-dependent NF-κB signaling pathway in cellular responses.

Formel: C₁₂H₁₁N₃O₂S
Lagerungstemperatur: Gefrierschrank
CAS-Nummer: 354811-10-2

Bestellen

Spezifikation Testergebnisse

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR