LY2409881 ≥98%
Lieferant: Cayman Chemical
|
Achtung
|
Synonyme:
2-(5-Chloro-2-((3-(4-methylpiperazin-1-yl)propyl)amino)pyrimidin-4-yl)-N-cyclopropylbenzo[b]thiophene-4-carboxamide trihydrochloride, 2-[5-chloro-2-[[3-(4-methyl-1-piperazinyl)propyl]amino]-4-pyrimidinyl]-N-cyclopropyl-benzo[b]thiophene-4-carboxamide, trihydrochloride
- A selective IKK2 inhibitor
A selective inhibitor of IKK2 (IC₅₀= 30 nm); suppresses constitutively activated NF-κB and induces apoptosis in lymphoma cells both in vitro and in preclinical mouse models of B and T cell lymphoma.
Formel:
C₂₄H₂₉ClN₆OS·₃HCl Molekulargewicht: 594.4 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD28900676 CAS-Nummer: 946518-60-1 UN-Nummer: 3077 ADR: 9 III |
Spezifikation Testergebnisse
Formulation | A crystalline solid |
Purity | ≥98% |
Stability | ≥ 2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...