5-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide ≥95%
Lieferant: Apollo Scientific
PC55719-250MG
PC55719-1G
APOSPC55719-250MGEA
248
EUR
APOSPC55719-250MG
APOSPC55719-1G
5-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide ≥95%
5-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide
Formel:
C₉H₄Br₂F₄O |
MDL Number:
MFCD32899475 CAS-Nummer: 2168316-92-3 UN-Nummer: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...