4-Bromo-1-(difluoromethyl)-2-iodobenzene ≥95%

Lieferant: Apollo Scientific

PC57120-250MG PC57120-1G
APOSPC57120-250MGEA 241 EUR
APOSPC57120-250MG APOSPC57120-1G
4-Bromo-1-(difluoromethyl)-2-iodobenzene ≥95%
4-Bromo-1-(difluoromethyl)-2-iodobenzene
Formel: C₇H₄BrF₂I
MDL Number: MFCD18391114
CAS-Nummer: 1261671-69-5
UN-Nummer: 3261

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR