5-Bromo-3-chloro-2-fluorophenacyl bromide 95%
Lieferant: Apollo Scientific
PC37884-250MG
PC37884-1G
APOSPC37884-250MGEA
219
EUR
APOSPC37884-250MG
APOSPC37884-1G
5-Bromo-3-chloro-2-fluorophenacyl bromide 95%
5-Bromo-3-chloro-2-fluorophenacyl bromide
Formel:
C₈H₄Br₂ClFO |
MDL Number:
MFCD28733998 CAS-Nummer: 1805518-57-3 UN-Nummer: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...