2-Bromo-6-fluoro-3-iodobenzaldehyde ≥95%

Lieferant: Apollo Scientific

PC56978-250MG PC56978-1G
APOSPC56978-250MGEA 261 EUR
APOSPC56978-250MG APOSPC56978-1G
2-Bromo-6-fluoro-3-iodobenzaldehyde ≥95%
2-Bromo-6-fluoro-3-iodobenzaldehyde
Formel: C₇H₃BrFIO
MDL Number: MFCD31660988
CAS-Nummer: 2092187-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR