Methyl 2-bromo-4-chloro-5-fluorobenzoate 95%
Lieferant: Apollo Scientific
PC57124-250MG
PC57124-1G
APOSPC57124-250MGEA
223
EUR
APOSPC57124-250MG
APOSPC57124-1G
Methyl 2-bromo-4-chloro-5-fluorobenzoate 95%
Methyl 2-bromo-4-chloro-5-fluorobenzoate
Formel:
C₈H₅BrClFO₂ |
MDL Number:
MFCD28733923 CAS-Nummer: 1807003-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...