2-Brom-4-chlor-5-fluoriodbenzol ≥95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-5-chloro-4-fluoro-2-iodobenzene

PC56008-250MG PC56008-1G PC56008-5G
APOSPC56008-250MGEA 55.1 EUR
APOSPC56008-250MG APOSPC56008-1G APOSPC56008-5G
2-Brom-4-chlor-5-fluoriodbenzol ≥95%
2-Brom-4-chlor-5-fluoriodbenzol
Formel: C₆H₂BrClFI
Molekulargewicht: 335.3400 g/mol
MDL Number: MFCD12026232
CAS-Nummer: 1067882-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR