5’-Bromo-2’-fluoro-3’-(trifluoromethyl)acetophenone ≥95%

Lieferant: Apollo Scientific

Synonyme: 1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)ethanone

PC55718-250MG PC55718-1G PC55718-5G
APOSPC55718-250MGEA 217 EUR
APOSPC55718-250MG APOSPC55718-1G APOSPC55718-5G
5’-Bromo-2’-fluoro-3’-(trifluoromethyl)acetophenone ≥95%
5’-Bromo-2’-fluoro-3’-(trifluoromethyl)acetophenone
Formel: C₉H₅BrF₄O
MDL Number: MFCD32899472
CAS-Nummer: 2092468-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR