2-Bromo-4-chloro-5-fluorobenzaldehyde ≥95%

Lieferant: Apollo Scientific
Achtung

PC53333-250MG PC53333-1G PC53333-5G
APOSPC53333-250MGEA 31.9 EUR
APOSPC53333-250MG APOSPC53333-1G APOSPC53333-5G
2-Bromo-4-chloro-5-fluorobenzaldehyde ≥95%
2-Bromo-4-chloro-5-fluorobenzaldehyde
Formel: C₇H₃BrClFO
MDL Number: MFCD20486524
CAS-Nummer: 1067882-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR