3,5-Dichloro-4-fluoro-2-iodoaniline ≥95%

Lieferant: Apollo Scientific

PC37881-250MG PC37881-1G PC37881-5G
APOSPC37881-250MGEA 219 EUR
APOSPC37881-250MG APOSPC37881-1G APOSPC37881-5G
3,5-Dichloro-4-fluoro-2-iodoaniline ≥95%
3,5-Dichloro-4-fluoro-2-iodoaniline
Formel: C₆H₃Cl₂FIN
MDL Number: MFCD29484812
CAS-Nummer: 1699384-74-1
UN-Nummer: 2811

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR