3-Fluoro-4-hydroxy-5-nitrobenzoic acid ≥95%

Lieferant: Apollo Scientific

PC502057-250MG PC502057-1G PC502057-5G
APOSPC502057-250MGEA 190 EUR
APOSPC502057-250MG APOSPC502057-1G APOSPC502057-5G
3-Fluoro-4-hydroxy-5-nitrobenzoic acid ≥95%
3-Fluoro-4-hydroxy-5-nitrobenzoic acid
Formel: C₇H₄FNO₅
Molekulargewicht: 201.10876 g/mol
MDL Number: MFCD15528841
CAS-Nummer: 1047621-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR