2-Bromo-3-chloro-5-iodobenzoic acid ≥94%
Lieferant: Apollo Scientific
OR61621-250MG
OR61621-1G
APOSOR61621-250MGEA
190
EUR
APOSOR61621-250MG
APOSOR61621-1G
2-Bromo-3-chloro-5-iodobenzoic acid ≥94%
2-Bromo-3-chloro-5-iodobenzoic acid
Formel:
C₇H₃BrClIO₂ |
CAS-Nummer:
875846-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...