(Bromoethynyl)triisopropylsilane ≥97%

Lieferant: Apollo Scientific

OR62151-1G OR62151-5G OR62151-10G
APOSOR62151-1GEA 43.5 EUR
APOSOR62151-1G APOSOR62151-5G APOSOR62151-10G
(Bromoethynyl)triisopropylsilane ≥97%
(Bromoethynyl)triisopropylsilane
Formel: C₁₁H₂₁BrSi
Molekulargewicht: 261.28 g/mol
Dichte: 1.09
MDL Number: MFCD11036302
CAS-Nummer: 111409-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR