5-Bromo-1-cyclopropyl-1H-imidazole ≥95%

Lieferant: Apollo Scientific

OR302570-250MG OR302570-1G
APOSOR302570-250MGEA 261 EUR
APOSOR302570-250MG APOSOR302570-1G
5-Bromo-1-cyclopropyl-1H-imidazole ≥95%
5-Bromo-1-cyclopropyl-1H-imidazole
Formel: C₆H₇BrN₂
Molekulargewicht: 187.03718 g/mol
MDL Number: MFCD16250307
CAS-Nummer: 1262035-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR