4,5-Difluoro-2-nitrobenzaldehyde ≥95%

Lieferant: Apollo Scientific

PC905972-1G PC905972-5G PC905972-10G
APOSPC905972-1GEA 290 EUR
APOSPC905972-1G APOSPC905972-5G APOSPC905972-10G
4,5-Difluoro-2-nitrobenzaldehyde ≥95%
4,5-Difluoro-2-nitrobenzaldehyde
Formel: C₇H₃F₂NO₃
MDL Number: MFCD28154587
CAS-Nummer: 149169-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR