5-Bromo-4-cyanoquinoline-n-oxide ≥95%

Lieferant: Apollo Scientific

OR70314-250MG OR70314-1G
APOSOR70314-250MGEA 227 EUR
APOSOR70314-250MG APOSOR70314-1G
5-Bromo-4-cyanoquinoline-n-oxide ≥95%
5-Bromo-4-cyanoquinoline-n-oxide
Formel: C₁₀H₅BrN₂O
MDL Number: MFCD32862642
CAS-Nummer: 1874272-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR