6-Bromo-4-cyanoquinoline-n-oxide ≥95%

Lieferant: Apollo Scientific

OR70313-250MG OR70313-1G
APOSOR70313-250MGEA 227 EUR
APOSOR70313-250MG APOSOR70313-1G
6-Bromo-4-cyanoquinoline-n-oxide ≥95%
6-Bromo-4-cyanoquinoline-n-oxide
Formel: C₁₀H₅BrN₂O
MDL Number: MFCD32862643
CAS-Nummer: 147689-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR