Phenol verflüssigt ≥89,0% USP
Lieferant: Spectrum Chemical
|
Gefahr
|
Stabilisierung:
20 ppm Stabilisiert mit Citronensäure
PH125-2.5LTPV
PH125-4LTGL
PH125-4LTPV
SPCMPH125-2.5LTPVEA
898
EUR
SPCMPH125-2.5LTPV
SPCMPH125-4LTGL
SPCMPH125-4LTPV
Phenol verflüssigt ≥89,0% USP
Phenol verflüssigt
Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₆H₅OH Molekulargewicht: 94.11 g/mol Dichte: 1.071 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002143 CAS-Nummer: 108-95-2 EINECS: 203-632-7 UN-Nummer: 2821 ADR: 6.1 II |
Spezifikation Testergebnisse
Assay | 89,0 % Min. |
Clarity of solution and reaction | Clear and neutral on acid to litmus |
Distilling range | 182.5°C Max. |
Nonvolatile residue | 0.05 % Max |
Elemental impurities | As reported |
Identification A | White precipitate forms |
Identification B | Violet color produced |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...