Nonoxynol-9
Lieferant: Spectrum Chemical
|
|
Nonoxynol 9 is also known as N-9 is used as a surfactant. It is an ingredient in several different types of cosmetic and cleaning products nonionic surfactants.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₃₃H₆₀O₁₀ Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
26571-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...