Buckets with Spout, Stainless Steel
Lieferant: Sampling Systems
A686-20 A686-15
SAMPA686-20EA 864 EUR
SAMPA686-20 SAMPA686-15
Buckets with Spout, Stainless Steel
Eimer
These buckets with spouts make emptying fast and simple.
  • Designed for heavy duty use
  • Perfect for use in pharma, chemical and food
  • Made from chemically resistant 316L stainless steel
  • Optional lid available
The Sampling Systems range of buckets with spouts are designed with a handy pouring spout which makes emptying fast and simple. Made from chemically resistant 316L stainless steel, they can be widely used across many applications in pharma, chemical and food.