N-(Phosphonomethyl)glycine

Lieferant: Thermo Fisher Scientific
Gefahr

466180010 466180050
ACRO466180010EA 67.8 EUR
ACRO466180010 ACRO466180050
N-(Phosphonomethyl)glycine
N-(Phosphonomethyl)glycine
Formel: C₃H₈NO₅P
Schmelzpunkt: 230°C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00055350
CAS-Nummer: 1071-83-6
EINECS: 213-997-4
UN-Nummer: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥ 94%
Water ≤ 1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR