C22 Phytoceramide-d₃ (t18:0/22:0-d₃) ≥99% deuterierte Formen (d1-d3)
Lieferant: Cayman Chemical
Synonyme:
Cer(t18:0/22:0-d3), N-Docosanoyl Phytosphingosine-d3, C22 Phytosphingosine-d3 (t18:0/22:0-d3), N-[(1S)-2S,3R-dihydroxy-1-(hydroxymethyl)heptadecyl]-docosanamide-d3, Ceramide (t18:0/22:0-d3), C22 Phytoceramide-d₃ (t18:0/22:0-d₃)
C22 Phytoceramide-d3 is intended for use as an internal standard for the quantification of C22 phytoceramide.
Bitte beachten This product is not for human or veterinary use.
Formel:
C₄₀H₇₈D₃NO₄ Lagerungstemperatur: Gefrierschrank |
CAS-Nummer:
2011762-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...