Double temperature probes, BOLA
Lieferant: Bohlender
P1740-20 P1740-23 P1740-30 P1740-40
620-1798EA 445 EUR
620-1798 620-1799 620-1800 620-1801
Double temperature probes, BOLA
Temperaturfühler
These temperature probe with two independent thermocouples in one PTFE-encapsulated stainless steel tube (1,4571) for parallel temperature measurement in aggressive liquids.
  • Temperature resistance from –200 to +250 °C; universal chemical resistance
  • With 2 PT100 thermocouples
  • Ideal for built-in measurements cables
With collar ring 12 mm Ø. Connection by two couplings (type Lemo®, socket size 1,4-pole) fixed directly at the end of the probe. Double safety due to redundant systems. Control function due to two independent thermocouples. Simultaneous temperature measurement and safety switching in only one port.